siRNA Details
Gene Symbol: HMGCLL1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001042406.1 | NP_001035865.1 | 2, 3 | 356 | ||
NM_001287741.1 | NP_001274670.1 | 2, 3 | 356 | |||
NM_001287746.1 | NP_001274675.1 | 3, 4 | 452 | |||
NM_001287753.1 | NP_001274682.1 | 2, 3 | 356 | |||
NM_019036.2 | NP_061909.2 | 3, 4 | 446 | |||
NR_109867.1 | 2, 3 | 356 | ||||
NR_109869.1 | 2, 3 | 356 | ||||
XM_011514700.2 | XP_011513002.1 | 2, 3 | 356 | |||
GenBank mRNA | AK055075.1 | - | 3, 4 | 337 | ||
AK127587.1 | - | 3, 4 | 419 | |||
AK294210.1 | - | 2, 3 | 308 | |||
AK297191.1 | - | 2, 3 | 247 | |||
AL137605.1 | - | 2, 3 | 215 | |||
BC024194.2 | - | 2, 3 | 237 | 4818781 | ||
DA508472.1 | - | 2, 3 | 355 | |||
DB179645.1 | - | 2, 3 | 357 |

More Information
Additional Information:
This siRNA no longer meets our design criteria. We suggest you choose a different siRNA for this gene or contact Tech Support for more information.
Gene Ontology Categories:
