siRNA Details
Gene Symbol: RBMX | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_002139.3 | NP_002130.2 | 9 | 1831 | ||
NR_028476.1 | 8 | 1659 | ||||
NR_028477.1 | 9 | 1866 | ||||
GenBank mRNA | AK091520.1 | - | 8 | 1605 | ||
AK096015.1 | - | 2 | 1737 | |||
BC006550.2 | - | 9 | 1767 | 3010123 | ||
BC007435.2 | - | 9 | 1767 | 3010123 | ||
BX647131.1 | - | 7 | 2235 | |||
Z23064.1 | - | 8 | 1630 |

More Information
Other Species Targeted

This siRNA also targets the following transcripts in Mouse: NM_011252, NM_001166623, AK135164, AK083899, AJ237847, and others.
Gene Ontology Categories:
