siRNA Details
Gene Symbol: ZNF563 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_145276.2 | NP_660319.1 | 4 | 820 | ||
XM_005259751.3 | XP_005259808.1 | 4 | 820 | |||
XM_006722650.3 | XP_006722713.1 | 3 | 757 | |||
XM_006722651.3 | XP_006722714.1 | 4 | 650 | |||
XM_011527698.1 | XP_011526000.1 | 4 | 640 | |||
XM_011527699.2 | XP_011526001.1 | 3 | 824 | |||
XM_011527700.2 | XP_011526002.1 | 3 | 595 | |||
XM_017026332.1 | XP_016881821.1 | 3 | 556 | |||
GenBank mRNA | AK092116.1 | - | 4 | 820 | ||
AK313755.1 | - | 4 | 724 | |||
BC022523.1 | - | 4 | 788 | 4794529 |

More Information
Additional Information:
This siRNA no longer meets our design criteria. We suggest you choose a different siRNA for this gene or contact Tech Support for more information.
Gene Ontology Categories:
