siRNA Details
Gene Symbol: TVP23C-CDRT4 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001204478.1 | NP_001191407.1 | 7 | 801 | ||
NR_037924.1 | 6 | 656 | ||||
GenBank mRNA | AB074181.1 | - | 7 | 731 | ||
AK303475.1 | - | 7 | 770 | |||
DA796743.1 | - | 6 | 497 |
Gene Symbol: CDRT4 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001204477.1 | NP_001191406.1 | 4 | 390 | ||
GenBank mRNA | AK093993.1 | - | 2 | 134 | ||
BC029542.1 | - | 4 | 390 | 5298701 | ||
BX379636.2 | - | 2 | 157 |

More Information
Additional Information:
This siRNA no longer meets our design criteria. We suggest you choose a different siRNA for this gene or contact Tech Support for more information.
