siRNA Details
Gene Symbol: LINC00301 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NR_026946.1 | 4 | 379 | |||
GenBank mRNA | AK058123.1 | - | 4 | 379 | ||
BC029583.1 | - | 4 | 357 | |||
BX103095.1 | - | 4 | 360 |

More Information
Additional Information:
This siRNA no longer meets our design criteria. We suggest you choose a different siRNA for this gene or contact Tech Support for more information.
