siRNA Details
Gene Symbol: HIST1H2AK | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_003510.2 | NP_003501.1 | 1 | 50 | ||
GenBank mRNA | AK311941.1 | - | 1 | 83 | ||
BC034487.2 | - | 1 | 86 | 4825623 | ||
BC104198.1 | - | 1 | 50 | 40031339 | ||
BC104199.1 | - | 1 | 50 | 40031340 |

More Information
Additional Information:
This siRNA no longer meets our design criteria. We suggest you choose a different siRNA for this gene or contact Tech Support for more information.
Gene Ontology Categories:
