siRNA Details
Gene Symbol: OR4C12 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001005270.4 | NP_001005270.3 | 1 | 915 | ||
GenBank mRNA | BC136850.1 | - | 1 | 915 | 9020841 | |
BC136851.1 | - | 1 | 915 | 9020842 |

More Information
Additional Information:
This siRNA no longer meets our design criteria. We suggest you choose a different siRNA for this gene or contact Tech Support for more information.
Gene Ontology Categories:
