siRNA Details
Gene Symbol: LOC100506990 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NR_040091.1 | 7 | 3187 | |||
GenBank mRNA | BC037934.1 | - | 7 | 3069 |

More Information
Additional Information:
This siRNA no longer meets our design criteria. We suggest you choose a different siRNA for this gene or contact Tech Support for more information.
