siRNA Details
Gene Symbol: KCNA4 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_002233.3 | NP_002224.1 | 2 | 1861 | ||
GenBank mRNA | AK313662.1 | - | 1 | 1140 | ||
L02751.1 | - | 1 | 620 | |||
M55514.1 | - | 2 | 1774 | |||
M60450.1 | - | 1 | 917 |

More Information
Additional Information:
This siRNA no longer meets our design criteria. We suggest you choose a different siRNA for this gene or contact Tech Support for more information.
Gene Ontology Categories:
