siRNA Details
Gene Symbol: KCNA4 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_002233.3 | NP_002224.1 | 2 | 2433 | ||
GenBank mRNA | AK313662.1 | - | 1 | 1712 | ||
L02751.1 | - | 1 | 1192 | |||
M55514.1 | - | 2 | 2346 | |||
M60450.1 | - | 1 | 1489 |

More Information
Other Species Targeted

Gene Ontology Categories:
