siRNA Details
Gene Symbol: FLT3 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_004119.2 | NP_004110.2 | 5 | 601 | ||
NR_130706.1 | 5 | 601 | ||||
XM_011535015.2 | XP_011533317.1 | 5 | 600 | |||
XM_011535017.2 | XP_011533319.1 | 5 | 503 | |||
XM_011535018.2 | XP_011533320.1 | 2 | 111 | |||
XM_017020486.1 | XP_016875975.1 | 5 | 596 | |||
XM_017020487.1 | XP_016875976.1 | 6 | 674 | |||
GenBank mRNA | BC036028.1 | - | 5 | 601 | ||
BC126350.1 | - | 5 | 585 | 8992066 | ||
BC144039.1 | - | 5 | 586 | 9052559 | ||
BC144040.1 | - | 5 | 586 | 9052560 | ||
BI461248.1 | - | 5 | 606 | |||
U02687.1 | - | 5 | 576 | |||
Z26652.1 | - | 5 | 519 |

More Information
Additional Information:
This siRNA no longer meets our design criteria. We suggest you choose a different siRNA for this gene or contact Tech Support for more information.
Gene Ontology Categories:
