siRNA Details
Gene Symbol: FLT3 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_004119.2 | NP_004110.2 | 7 | 854 | ||
NR_130706.1 | 7 | 854 | ||||
XM_011535015.2 | XP_011533317.1 | 7 | 853 | |||
XM_011535017.2 | XP_011533319.1 | 7 | 756 | |||
XM_011535018.2 | XP_011533320.1 | 4 | 364 | |||
XM_017020487.1 | XP_016875976.1 | 8 | 927 | |||
XM_017020488.1 | XP_016875977.1 | 6 | 696 | |||
XM_017020489.1 | XP_016875978.1 | 6 | 715 | |||
GenBank mRNA | BC036028.1 | - | 7 | 854 | ||
BC126350.1 | - | 7 | 838 | 8992066 | ||
BC144039.1 | - | 7 | 839 | 9052559 | ||
BC144040.1 | - | 7 | 839 | 9052560 | ||
U02687.1 | - | 7 | 829 | |||
Z26652.1 | - | 7 | 772 |

More Information
Additional Information:
This siRNA no longer meets our design criteria. We suggest you choose a different siRNA for this gene or contact Tech Support for more information.
Gene Ontology Categories:
