siRNA Details
Gene Symbol: LYSMD3 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001286812.1 | NP_001273741.1 | 3 | 1665 | ||
NM_198273.2 | NP_938014.1 | 3 | 1724 | |||
XM_005248421.4 | XP_005248478.1 | 3 | 2189 | |||
GenBank mRNA | AK023604.1 | - | 1 | 460 | ||
AL137315.1 | - | 1 | 1226 | |||
AL832721.1 | - | 3 | 1705 | |||
BC136740.1 | - | 3 | 1621 | 9020730 | ||
BC136741.1 | - | 3 | 1616 | 9020731 | ||
BX537972.1 | - | 3 | 1699 | |||
CR933624.1 | - | 4 | 1315 |
