siRNA Details
Gene Symbol: LINC00934 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NR_024246.2 | 3, 4 | 1441 | |||
GenBank mRNA | AK057497.1 | - | 3, 4 | 1441 | ||
BC118632.1 | - | 1, 2 | 358 |

UserName
Please enter the information below and press OK to send your cart to Core Services for purchase.
Gene Symbol: LINC00934 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NR_024246.2 | 3, 4 | 1441 | |||
GenBank mRNA | AK057497.1 | - | 3, 4 | 1441 | ||
BC118632.1 | - | 1, 2 | 358 |