siRNA Details
Gene Symbol: OVAAL | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NR_125716.1 | 3 | 909 | |||
GenBank mRNA | BC036830.1 | - | 3 | 906 | ||
BC044260.1 | - | 3 | 907 | |||
BC063624.1 | - | 3 | 909 | |||
BC073892.1 | - | 3 | 933 | |||
BC107121.2 | - | 2 | 813 | |||
BC107122.1 | - | 2 | 813 |
