siRNA Details
Gene Symbol: C1orf112 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001320047.1 | NP_001306976.1 | 10, 11 | 1188 | ||
NM_001320048.1 | NP_001306977.1 | 8, 9 | 943 | |||
NM_018186.3 | NP_060656.2 | 9, 10 | 1491 | |||
XM_005245317.4 | XP_005245374.1 | 9, 10 | 1063 | |||
XM_011509735.2 | XP_011508037.1 | 6, 7 | 673 | |||
XM_017001722.1 | XP_016857211.1 | 9, 10 | 1063 | |||
XM_017001723.1 | XP_016857212.1 | 9, 10 | 1063 | |||
XM_017001724.1 | XP_016857213.1 | 7, 8 | 812 | |||
XM_017001725.1 | XP_016857214.1 | 6, 7 | 673 | |||
GenBank mRNA | AK001568.1 | - | 8, 9 | 863 | ||
AK096493.1 | - | 10, 11 | 1098 | |||
AK127098.1 | - | 13, 14 | 1885 | |||
AK294569.1 | - | 5, 6 | 634 | |||
AK299369.1 | - | 9, 10 | 974 | |||
AK303296.1 | - | 8, 9 | 939 | |||
AL354614.1 | - | 9, 10 | 1491 | |||
BC091516.1 | - | 10, 11 | 1102 | 30389801 | ||
BC107168.1 | - | 8, 9 | 840 | 40035373 | ||
BC107169.1 | - | 9, 10 | 979 | 40035375 |
