siRNA Details
Gene Symbol: SHISA3 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001080505.1 | NP_001073974.1 | 2 | 1058 | ||
GenBank mRNA | BC012029.1 | - | 1 | 558 |

UserName
Please enter the information below and press OK to send your cart to Core Services for purchase.
Gene Symbol: SHISA3 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001080505.1 | NP_001073974.1 | 2 | 1058 | ||
GenBank mRNA | BC012029.1 | - | 1 | 558 |