siRNA Details
Gene Symbol: LOC153910 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NR_027311.1 | 3 | 982 | |||
NR_027312.1 | 3 | 985 | ||||
GenBank mRNA | AK055933.1 | - | 3 | 985 | ||
BC033340.1 | - | 1 | 304 | |||
BC125150.1 | - | 3 | 982 |
