siRNA Details
Gene Symbol: ZNF846 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001077624.1 | NP_001071092.1 | 3, 4 | 684 | ||
XM_005259772.3 | XP_005259829.1 | 3, 4 | 475 | |||
XM_006722658.3 | XP_006722721.1 | 3, 4 | 684 | |||
XM_011527717.2 | XP_011526019.1 | 3, 4 | 487 | |||
XM_011527719.2 | XP_011526021.1 | 3, 4 | 503 | |||
XM_011527720.2 | XP_011526022.1 | 2, 3 | 258 | |||
XM_011527721.2 | XP_011526023.1 | 1, 2 | 32 | |||
XM_017026404.1 | XP_016881893.1 | 3, 4 | 642 | |||
XM_017026405.1 | XP_016881894.1 | 3, 4 | 1207 | |||
XM_017026407.1 | XP_016881896.1 | 3, 4 | 684 | |||
GenBank mRNA | AK308938.1 | - | 3, 4 | 567 | ||
BC118576.1 | - | 3, 4 | 255 | 40053684 | ||
BC118657.1 | - | 3, 4 | 255 |
