siRNA Details
Gene Symbol: C10orf128 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001010863.2 | NP_001010863.1 | 6 | 434 | ||
NM_001288740.1 | NP_001275669.1 | 7 | 513 | |||
NM_001288741.1 | NP_001275670.1 | 5 | 403 | |||
NM_001288742.1 | NP_001275671.1 | 5 | 753 | |||
NM_001288743.1 | NP_001275672.1 | 7 | 941 | |||
XM_011539379.1 | XP_011537681.1 | 7 | 791 | |||
XM_011539380.1 | XP_011537682.1 | 7 | 771 | |||
XM_011539383.2 | XP_011537685.1 | 6 | 964 | |||
XM_011539384.1 | XP_011537686.1 | 6 | 634 | |||
XM_011539385.1 | XP_011537687.1 | 5 | 603 | |||
GenBank mRNA | AL524299.3 | - | 7 | 488 | ||
AL832399.1 | - | 1 | 816 | |||
BC031641.1 | - | 6 | 965 | |||
BC047724.1 | - | 5 | 1462 | |||
BU145920.1 | - | 5 | 665 | |||
DA366138.1 | - | 5 | 356 |
