siRNA Details
Gene Symbol: C10orf128 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001288742.1 | NP_001275671.1 | 4, 5 | 334 | ||
NM_001288743.1 | NP_001275672.1 | 6, 7 | 522 | |||
XM_011539383.2 | XP_011537685.1 | 5, 6 | 545 | |||
GenBank mRNA | BU145920.1 | - | 4, 5 | 246 | ||
DQ884399.1 | - | 6, 7 | 458 |
