siRNA Details
Gene Symbol: FRMD4B | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_015123.2 | NP_055938.1 | 19 | 2183 | ||
XM_005264722.1 | XP_005264779.1 | 19 | 1860 | |||
XM_005264723.1 | XP_005264780.1 | 9 | 1342 | |||
XM_017005989.1 | XP_016861478.1 | 19 | 1967 | |||
XM_017005990.1 | XP_016861479.1 | 19 | 1860 | |||
XM_017005991.1 | XP_016861480.1 | 20 | 2020 | |||
XM_017005992.1 | XP_016861481.1 | 19 | 1800 | |||
XM_017005993.1 | XP_016861482.1 | 19 | 1791 | |||
XM_017005994.1 | XP_016861483.1 | 19 | 3633 | |||
XM_017005995.1 | XP_016861484.1 | 13 | 1377 | |||
XM_017005996.1 | XP_016861485.1 | 9 | 1079 | |||
XM_017005997.1 | XP_016861486.1 | 9 | 1342 | |||
GenBank mRNA | AB023230.1 | - | 19 | 1952 | ||
AK024086.1 | - | 3 | 218 | |||
AK295045.1 | - | 14 | 1425 | |||
AL832231.1 | - | 9 | 1180 | |||
BC110637.1 | - | 4 | 404 |
