siRNA Details
Gene Symbol: TSR1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_018128.4 | NP_060598.3 | 6, 7 | 2105 | ||
GenBank mRNA | AB037822.1 | - | 6, 7 | 2228 | ||
AK001396.1 | - | 4, 5 | 747 | |||
BC110851.1 | - | 6, 7 | 1683 | 5786818 | ||
BC126110.1 | - | 6, 7 | 1653 | 8991826 |
