siRNA Details
Gene Symbol: THSD7A | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_015204.2 | NP_056019.1 | 7 | 2229 | ||
XM_006715659.1 | XP_006715722.1 | 7 | 2228 | |||
XM_006715660.1 | XP_006715723.1 | 7 | 2228 | |||
XM_006715662.1 | XP_006715725.1 | 7 | 2228 | |||
XM_011515193.2 | XP_011513495.1 | 7 | 4553 | |||
XM_011515194.1 | XP_011513496.1 | 7 | 1994 | |||
GenBank mRNA | AB023177.2 | - | 6 | 1513 | ||
AK092252.1 | - | 7 | 2229 |
