siRNA Details
Gene Symbol: LINC00710 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NR_015413.1 | 5 | 571 | |||
NR_024410.1 | 4 | 501 | ||||
NR_024411.1 | 3 | 390 | ||||
GenBank mRNA | BC033338.1 | - | 5 | 503 | ||
BC106956.2 | - | 4 | 305 | |||
BC106957.2 | - | 3 | 194 | |||
BC113009.1 | - | 3 | 194 |
