siRNA Details
Gene Symbol: LOC255187 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NR_122044.1 | 2 | 439 | |||
GenBank mRNA | AK056780.1 | - | 2 | 436 | ||
BG178577.1 | - | 3 | 547 | |||
BX106527.1 | - | 2 | 301 |

UserName
Please enter the information below and press OK to send your cart to Core Services for purchase.
Gene Symbol: LOC255187 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NR_122044.1 | 2 | 439 | |||
GenBank mRNA | AK056780.1 | - | 2 | 436 | ||
BG178577.1 | - | 3 | 547 | |||
BX106527.1 | - | 2 | 301 |