siRNA Details
Gene Symbol: LINC00935 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NR_029448.1 | 4, 5 | 474 | |||
NR_029449.1 | 3, 4 | 389 | ||||
GenBank mRNA | AK131005.1 | - | 3, 4 | 389 | ||
BC036043.1 | - | 3, 4 | 364 | |||
BC054859.1 | - | 4, 5 | 456 | |||
BC146870.1 | - | 3, 4 | 371 | |||
BC146877.1 | - | 3, 4 | 371 |
