siRNA Details
Gene Symbol: HSPA12A | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_025015.2 | NP_079291.2 | 12 | 3774 | ||
XM_005269672.3 | XP_005269729.1 | 13 | 3924 | |||
XM_005269673.4 | XP_005269730.1 | 13 | 4868 | |||
XM_011539579.2 | XP_011537881.1 | 14 | 3985 | |||
XM_011539580.2 | XP_011537882.1 | 13 | 3930 | |||
XM_017016032.1 | XP_016871521.1 | 10 | 3496 | |||
GenBank mRNA | AB007877.1 | - | 12 | 3774 |
