siRNA Details
Gene Symbol: BTBD7 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_018167.4 | NP_060637.1 | 4 | 1827 | ||
GenBank mRNA | AK001510.1 | - | 4 | 1753 | ||
BC047071.1 | - | 4 | 1824 | |||
BC141850.1 | - | 4 | 1768 |

More Information
Additional Information:
This siRNA no longer meets our design criteria. We suggest you choose a different siRNA for this gene or contact Tech Support for more information.
Gene Ontology Categories:
