siRNA Details
Gene Symbol: HCCAT5 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NR_027756.1 | 3 | 983 | |||
GenBank mRNA | FJ222407.1 | - | 3 | 983 | ||
FJ222408.1 | - | 1 | 1330 |

UserName
Please enter the information below and press OK to send your cart to Core Services for purchase.
Gene Symbol: HCCAT5 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NR_027756.1 | 3 | 983 | |||
GenBank mRNA | FJ222407.1 | - | 3 | 983 | ||
FJ222408.1 | - | 1 | 1330 |