siRNA Details
Gene Symbol: TMEM235 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001204210.1 | NP_001191139.1 | 6 | 2342 | ||
NM_001204211.1 | NP_001191140.1 | 5 | 2261 | |||
NM_001204212.1 | NP_001191141.1 | 4 | 2123 | |||
XM_011524620.2 | XP_011522922.1 | 7 | 1869 | |||
XM_011524624.2 | XP_011522926.1 | 5 | 1707 | |||
XM_017024459.1 | XP_016879948.1 | 6 | 1782 | |||
XM_017024460.1 | XP_016879949.1 | 7 | 1880 | |||
GenBank mRNA | BC042066.1 | - | 4 | 2123 |
