siRNA Details
Gene Symbol: TMEM235 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001204210.1 | NP_001191139.1 | 2 | 715 | ||
NM_001204211.1 | NP_001191140.1 | 2 | 715 | |||
NM_001204212.1 | NP_001191141.1 | 2 | 715 | |||
XM_011524620.2 | XP_011522922.1 | 1 | 91 | |||
XM_011524624.2 | XP_011522926.1 | 1 | 91 | |||
XM_017024459.1 | XP_016879948.1 | 1 | 91 | |||
XM_017024460.1 | XP_016879949.1 | 1 | 91 | |||
XM_017024461.1 | XP_016879950.1 | 1 | 91 | |||
GenBank mRNA | BC042066.1 | - | 2 | 715 | ||
BI757060.1 | - | 1 | 53 |

More Information
Other Species Targeted

