siRNA Details
Gene Symbol: ZNF841 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001136499.1 | NP_001129971.1 | 6, 7 | 727 | ||
XM_011526833.2 | XP_011525135.1 | 5, 6 | 619 | |||
XM_017026652.1 | XP_016882141.1 | 2, 3 | 276 | |||
XM_017026653.1 | XP_016882142.1 | 6, 7 | 685 | |||
XM_017026654.1 | XP_016882143.1 | 6, 7 | 671 | |||
XM_017026655.1 | XP_016882144.1 | 2, 3 | 273 | |||
XM_017026656.1 | XP_016882145.1 | 5, 6 | 2322 | |||
XM_017026657.1 | XP_016882146.1 | 5, 6 | 616 | |||
XM_017026658.1 | XP_016882147.1 | 2, 3 | 276 | |||
XM_017026659.1 | XP_016882148.1 | 2, 3 | 276 | |||
XM_017026660.1 | XP_016882149.1 | 2, 3 | 276 | |||
XM_017026661.1 | XP_016882150.1 | 2, 3 | 276 | |||
XM_017026662.1 | XP_016882151.1 | 5, 6 | 2322 | |||
XM_017026663.1 | XP_016882152.1 | 5, 6 | 462 | |||
GenBank mRNA | BC136254.1 | - | 3, 4 | 317 | 9020241 |
