siRNA Details
Gene Symbol: ZNF841 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001136499.1 | NP_001129971.1 | 7 | 3069 | ||
NM_001321349.1 | NP_001308278.1 | 5 | 2813 | |||
XM_011526833.2 | XP_011525135.1 | 6 | 2961 | |||
XM_017026652.1 | XP_016882141.1 | 3 | 2618 | |||
XM_017026653.1 | XP_016882142.1 | 7 | 3027 | |||
XM_017026654.1 | XP_016882143.1 | 7 | 3013 | |||
XM_017026655.1 | XP_016882144.1 | 3 | 2615 | |||
XM_017026656.1 | XP_016882145.1 | 6 | 4664 | |||
XM_017026657.1 | XP_016882146.1 | 6 | 2958 | |||
GenBank mRNA | AK093512.1 | - | 1 | 1550 | ||
AK094953.1 | - | 1 | 1174 | |||
AK129938.1 | - | 2 | 1372 | |||
AK131409.1 | - | 5 | 2813 | |||
BC136254.1 | - | 4 | 2659 | 9020241 |
