siRNA Details
Gene Symbol: SBNO1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001167856.1 | NP_001161328.1 | 31 | 5066 | ||
NM_018183.3 | NP_060653.3 | 31 | 5063 | |||
XM_005253572.3 | XP_005253629.1 | 32 | 5204 | |||
XM_005253573.4 | XP_005253630.1 | 33 | 5614 | |||
XM_005253575.3 | XP_005253632.1 | 32 | 5201 | |||
XM_005253576.3 | XP_005253633.1 | 33 | 5245 | |||
XM_006719473.3 | XP_006719536.1 | 34 | 5370 | |||
XM_006719474.3 | XP_006719537.1 | 33 | 5323 | |||
XM_011538533.2 | XP_011536835.1 | 34 | 5364 | |||
XM_011538534.2 | XP_011536836.1 | 31 | 5102 | |||
XM_011538535.2 | XP_011536837.1 | 31 | 5104 | |||
XM_017019556.1 | XP_016875045.1 | 32 | 5188 | |||
XM_017019557.1 | XP_016875046.1 | 32 | 5195 | |||
XM_017019558.1 | XP_016875047.1 | 33 | 5258 | |||
XM_017019559.1 | XP_016875048.1 | 32 | 5325 | |||
GenBank mRNA | AK001563.1 | - | 8 | 1809 | ||
AK074256.1 | - | 15 | 2634 | |||
AK096864.1 | - | 14 | 2436 |

More Information
Additional Information:
This siRNA no longer meets our design criteria. We suggest you choose a different siRNA for this gene or contact Tech Support for more information.
Gene Ontology Categories:
