siRNA Details
Gene Symbol: C1orf112 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001320047.1 | NP_001306976.1 | 24 | 2700 | ||
NM_001320048.1 | NP_001306977.1 | 22 | 2455 | |||
NM_018186.3 | NP_060656.2 | 23 | 3003 | |||
XM_005245317.4 | XP_005245374.1 | 23 | 2575 | |||
XM_011509735.2 | XP_011508037.1 | 20 | 2185 | |||
XM_017001722.1 | XP_016857211.1 | 23 | 2575 | |||
XM_017001724.1 | XP_016857213.1 | 22 | 2447 | |||
XM_017001725.1 | XP_016857214.1 | 21 | 2308 | |||
GenBank mRNA | AK001568.1 | - | 22 | 2375 | ||
AK023532.1 | - | 13 | 1438 | |||
AK096493.1 | - | 24 | 2610 | |||
AK127098.1 | - | 28 | 3492 | |||
AK294569.1 | - | 19 | 2146 | |||
AK310638.1 | - | 19 | 2116 | |||
AL354614.1 | - | 23 | 3003 | |||
BC091516.1 | - | 24 | 2614 | 30389801 | ||
BC107168.1 | - | 22 | 2352 | 40035373 | ||
BC107169.1 | - | 23 | 2491 | 40035375 |
