siRNA Details
Gene Symbol: TMLHE | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_018196.3 | NP_060666.1 | 8 | 2608 | ||
XM_011531182.2 | XP_011529484.1 | 8 | 2476 | |||
GenBank mRNA | AK001589.1 | - | 8 | 2583 |

More Information
Additional Information:
This siRNA no longer meets our design criteria. We suggest you choose a different siRNA for this gene or contact Tech Support for more information.
Gene Ontology Categories:
