siRNA Details
Gene Symbol: ZFP64 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_018197.2 | NP_060667.2 | 6 | 3031 | ||
NM_022088.4 | NP_071371.3 | 5 | 2869 | |||
NM_199426.1 | NP_955458.1 | 6 | 3025 | |||
XM_005260449.1 | XP_005260506.1 | 5 | 2448 | |||
XM_011528900.1 | XP_011527202.1 | 7 | 2922 | |||
XM_017027946.1 | XP_016883435.1 | 5 | 2591 | |||
XM_017027948.1 | XP_016883437.1 | 6 | 2570 | |||
XM_017027949.1 | XP_016883438.1 | 7 | 2889 | |||
GenBank mRNA | AK001596.1 | - | 6 | 2820 | ||
AK090547.1 | - | 7 | 2693 | |||
BC012759.2 | - | 6 | 3030 | 3634346 | ||
BC041622.1 | - | 6 | 3030 | 3633938 |

More Information
Additional Information:
This siRNA no longer meets our design criteria. We suggest you choose a different siRNA for this gene or contact Tech Support for more information.
Gene Ontology Categories:
