siRNA Details
Gene Symbol: DNAJC11 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_018198.3 | NP_060668.2 | 16 | 1912 | ||
XM_017001745.1 | XP_016857234.1 | 15 | 1765 | |||
GenBank mRNA | AF306695.1 | - | 16 | 1826 | ||
AK001599.1 | - | 16 | 1821 | |||
AK025491.1 | - | 6 | 587 | |||
AK093709.1 | - | 4 | 1067 | |||
AK095386.1 | - | 7 | 1634 | |||
AK294539.1 | - | 15 | 1735 | |||
AK298378.1 | - | 16 | 1798 | |||
AL833841.1 | - | 16 | 1786 | |||
AY336749.1 | - | 16 | 1708 | |||
BC006086.1 | - | 16 | 1808 | 3946414 | ||
BC008772.1 | - | 16 | 1813 | |||
BC014145.2 | - | 15 | 1664 | 4762069 |

More Information
Additional Information:
This siRNA no longer meets our design criteria. We suggest you choose a different siRNA for this gene or contact Tech Support for more information.
Gene Ontology Categories:
