siRNA Details
Gene Symbol: SMLR1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001195597.1 | NP_001182526.1 | 2 | 1906 | ||
GenBank mRNA | AK091900.1 | - | 2 | 1905 |

UserName
Please enter the information below and press OK to send your cart to Core Services for purchase.
Gene Symbol: SMLR1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001195597.1 | NP_001182526.1 | 2 | 1906 | ||
GenBank mRNA | AK091900.1 | - | 2 | 1905 |