siRNA Details
Gene Symbol: CNPY1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001103176.1 | NP_001096646.1 | 4 | 1658 | ||
GenBank mRNA | AK094445.1 | - | 4 | 1676 | ||
BC105732.1 | - | 4 | 1659 | |||
BC105733.1 | - | 4 | 1659 | |||
BC105734.1 | - | 4 | 1659 | |||
BC107598.1 | - | 4 | 1659 |
