siRNA Details
Gene Symbol: PNMAL1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001103149.1 | NP_001096619.1 | 3 | 2232 | ||
NM_018215.3 | NP_060685.2 | 3 | 2437 | |||
XM_011527067.2 | XP_011525369.1 | 3 | 2522 | |||
GenBank mRNA | AK001643.1 | - | 2 | 1456 | ||
AK096326.1 | - | 3 | 1997 | |||
AL834344.1 | - | 3 | 2070 | |||
BC026026.1 | - | 2 | 1470 | |||
BC032508.1 | - | 3 | 2415 | 5246377 | ||
BC051688.1 | - | 3 | 2394 | 5295669 | ||
BU844519.1 | - | 1 | 427 | |||
BX649091.1 | - | 2 | 1269 |
