siRNA Details
Gene Symbol: LOC286238 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001100111.1 | NP_001093581.1 | 2 | 651 | ||
GenBank mRNA | AK098532.1 | - | 2 | 629 | ||
DB454766.2 | - | 2 | 651 | |||
DB520173.2 | - | 1 | 162 |
