siRNA Details
Gene Symbol: LINC00705 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NR_015425.1 | 3 | 680 | |||
GenBank mRNA | BC040895.1 | - | 4 | 1409 | ||
BC101460.1 | - | 4 | 680 | |||
BC101989.1 | - | 4 | 680 | |||
BC104742.1 | - | 3 | 680 | |||
BC129979.1 | - | 4 | 680 |
