siRNA Details
Gene Symbol: C17orf51 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001113434.4 | NP_001106905.1 | 2 | 3278 | ||
GenBank mRNA | AK023039.1 | - | 1 | 2586 | ||
AK056436.1 | - | 2 | 3244 | |||
AK090937.1 | - | 4 | 1927 | |||
BC010612.1 | - | 2 | 3276 | |||
BC019938.1 | - | 2 | 3266 | |||
BC040371.1 | - | 1 | 2090 | |||
BC052777.1 | - | 2 | 3277 | |||
BC053952.1 | - | 2 | 3277 | |||
BC132738.1 | - | 2 | 3252 | 40146760 | ||
BC136630.1 | - | 2 | 3252 | 9020620 | ||
BX640850.1 | - | 4 | 2313 |
