siRNA Details
Gene Symbol: C17orf51 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001113434.4 | NP_001106905.1 | 2 | 3439 | ||
GenBank mRNA | AK023039.1 | - | 1 | 2747 | ||
AK056436.1 | - | 2 | 3405 | |||
AK090937.1 | - | 4 | 2088 | |||
BC010612.1 | - | 2 | 3437 | |||
BC019938.1 | - | 2 | 3427 | |||
BC040371.1 | - | 1 | 2251 | |||
BC052777.1 | - | 2 | 3438 | |||
BC053952.1 | - | 2 | 3438 | |||
BC132738.1 | - | 2 | 3413 | 40146760 | ||
BC136630.1 | - | 2 | 3413 | 9020620 | ||
BX640850.1 | - | 4 | 2474 |
