siRNA Details
Gene Symbol: LOC102724279 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001302493.1 | NP_001289422.1 | 3 | 216 | ||
GenBank mRNA | BX095604.1 | - | 3 | 216 | ||
GD139371.1 | - | 3 | 191 |

UserName
Please enter the information below and press OK to send your cart to Core Services for purchase.
Gene Symbol: LOC102724279 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001302493.1 | NP_001289422.1 | 3 | 216 | ||
GenBank mRNA | BX095604.1 | - | 3 | 216 | ||
GD139371.1 | - | 3 | 191 |