siRNA Details
Gene Symbol: UQCC1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_018244.4 | NP_060714.3 | 4 | 448 | ||
NM_199487.2 | NP_955781.2 | 4 | 448 | |||
XM_011528877.1 | XP_011527179.1 | 5 | 395 | |||
XM_011528878.1 | XP_011527180.1 | 3 | 257 | |||
XM_011528879.1 | XP_011527181.1 | 4 | 299 | |||
XM_011528880.1 | XP_011527182.1 | 4 | 357 | |||
XM_011528881.2 | XP_011527183.1 | 3 | 379 | |||
XM_011528882.1 | XP_011527184.1 | 4 | 306 | |||
XM_011528883.1 | XP_011527185.1 | 3 | 232 | |||
XM_011528884.1 | XP_011527186.1 | 3 | 183 | |||
GenBank mRNA | AK001712.1 | - | 4 | 279 | ||
AK022650.1 | - | 4 | 279 | |||
AK022742.1 | - | 5 | 402 | |||
AK225281.1 | - | 4 | 288 | |||
AK225557.1 | - | 4 | 279 | |||
AK295379.1 | - | 2 | 124 | |||
AK300130.1 | - | 4 | 279 | |||
AK308819.1 | - | 4 | 296 | |||
AU280736.1 | - | 4 | 279 | |||
AY050659.1 | - | 4 | 448 | |||
AY050660.1 | - | 3 | 251 | |||
BC105791.1 | - | 4 | 283 | 6458054 | ||
BC119013.1 | - | 4 | 314 | 40085306 | ||
BC122862.1 | - | 4 | 314 | 40085305 | ||
BM919736.1 | - | 4 | 301 |

More Information
Additional Information:
This siRNA no longer meets our design criteria. We suggest you choose a different siRNA for this gene or contact Tech Support for more information.
Gene Ontology Categories:
