siRNA Details
Gene Symbol: MROH2A | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001287395.1 | NP_001274324.1 | 38 | 4614 | ||
XM_011511075.2 | XP_011509377.1 | 38 | 4821 | |||
XM_011511076.2 | XP_011509378.1 | 38 | 4809 | |||
XM_011511077.2 | XP_011509379.1 | 38 | 4791 | |||
XM_011511078.2 | XP_011509380.1 | 38 | 4782 | |||
XM_011511079.2 | XP_011509381.1 | 38 | 4776 | |||
XM_011511080.2 | XP_011509382.1 | 38 | 6292 | |||
XM_011511081.2 | XP_011509383.1 | 37 | 4716 | |||
XM_011511082.2 | XP_011509384.1 | 37 | 4683 | |||
XM_011511083.2 | XP_011509385.1 | 38 | 4821 | |||
XM_011511084.2 | XP_011509386.1 | 37 | 4623 | |||
XM_011511085.2 | XP_011509387.1 | 32 | 3891 | |||
XM_011511086.2 | XP_011509388.1 | 29 | 3546 | |||
GenBank mRNA | BC042133.1 | - | 3 | 441 |
