siRNA Details
Gene Symbol: LINC01115 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NR_033880.3 | 4 | 800 | |||
NR_111963.1 | 3 | 563 | ||||
GenBank mRNA | BC043553.1 | - | 4 | 801 | ||
BC128382.1 | - | 3 | 328 | |||
BC128532.1 | - | 3 | 328 |
